Assay Details
| Gene Symbol: Tas2r126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207028.1 | NP_996911.1 | 1 | 878 | 116 | |
| GenBank mRNA | BC126974.1 | - | 1 | 878 | 40101187 | 116 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111083 | NM_207028.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111083 | BC126974.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113054 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111083 | NM_207028.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111083 | BC126974.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24398348 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113054 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107688 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113054 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05485294 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05751004 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05485297 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04998915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24398331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05166589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05336189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05610045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05485296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05473841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05336190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05485288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05485286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05610047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05849791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05849795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05049234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05360474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24398368 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10537790 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10537789 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600003727.mm.1 | TC0600000463.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600003727.mm.1 | NM_207028.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600003727.mm.1 | NM_207028.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5270711 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5270711 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270711 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253212 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253212 | - | - |
| mouse-6 v1.1 (Illumina) | scl0387353.1_95-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270711 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0387353.1_95-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270711 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2798839 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2798839 | - | - |

Tas2r126
Genomic Map