Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000011426.mm.1 |
NM_001113199.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011426.mm.1 |
NM_001113199.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011426.mm.1 |
NM_001282976.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011426.mm.1 |
NM_013608.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011426.mm.1 |
NM_013608.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011426.mm.1 |
TC1000001580.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011422.mm.1 |
NM_001113199.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011422.mm.1 |
NM_001113199.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011422.mm.1 |
NM_001282976.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011422.mm.1 |
NM_013608.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011422.mm.1 |
NM_013608.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011422.mm.1 |
TC1000001580.mm.1 |
MTA TC |