Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100017513.mm.1 |
NM_001048057.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017513.mm.1 |
NM_001048057.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017513.mm.1 |
NM_001048058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017513.mm.1 |
NM_001048058.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017513.mm.1 |
NM_023372.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017513.mm.1 |
NM_023372.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017513.mm.1 |
TC1100001874.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017514.mm.1 |
NM_001048057.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017514.mm.1 |
NM_001048057.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017514.mm.1 |
NM_001048058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017514.mm.1 |
NM_001048058.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017514.mm.1 |
NM_023372.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017514.mm.1 |
NM_023372.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017514.mm.1 |
TC1100001874.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017505.mm.1 |
NM_001048057.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017505.mm.1 |
NM_001048057.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017505.mm.1 |
NM_023372.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017505.mm.1 |
NM_023372.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017505.mm.1 |
TC1100001874.mm.1 |
MTA TC |