Assay Details
| Gene Symbol: Mrpl48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198831.2 | NP_942128.2 | 1 - 2 | 145 | 153 | |
| GenBank mRNA | BU604260.1 | - | 1 - 2 | 138 | 153 | |
| DN174444.1 | - | 1 - 2 | 145 | 153 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_198831.2 transcripts.
Set Membership: |
|

Mrpl48
Genomic Map