Assay Details
| Gene Symbol: Mrps21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078479.3 | NP_510964.1 | 1 - 2 | 124 | 155 | |
| GenBank mRNA | AB049956.1 | - | 2 - 3 | 149 | 155 | |
| AK002588.1 | - | 2 - 3 | 152 | 155 | ||
| AK003462.1 | - | 2 - 3 | 145 | 155 | ||
| AK003602.1 | - | 2 - 3 | 144 | 155 | ||
| AK007662.1 | - | 2 - 3 | 153 | 155 | ||
| AK007838.1 | - | 1 - 2 | 135 | 155 | ||
| AK008265.1 | - | 2 - 3 | 167 | 155 | ||
| AK008293.1 | - | 2 - 3 | 146 | 155 | ||
| AK009049.1 | - | 2 - 3 | 145 | 155 | ||
| AK009703.1 | - | 2 - 3 | 153 | 155 | ||
| BC027546.1 | - | 1 - 2 | 128 | 1515797 | 155 | |
| BC125658.1 | - | 1 - 2 | 125 | 40130173 | 155 | |
| BC125660.1 | - | 1 - 2 | 125 | 40130175 | 155 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AB049956.1,AK002588.1,AK003462.1,AK003602.1,AK007662.1,AK007838.1,AK008265.1,AK008293.1,AK009049.1,AK009703.1,BC027546.1,BC125658.1,BC125660.1,NM_078479.3 transcripts.
This assay is in the 5'UTR region of the AB049956.1,AK002588.1,AK003462.1,AK003602.1,AK007662.1,AK007838.1,AK008265.1,AK008293.1,AK009049.1,AK009703.1,BC027546.1,BC125658.1,BC125660.1,NM_078479.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346101 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346103 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346099 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346100 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346102 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126063 | AK003602.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126063 | NM_078479.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126063 | AK007662.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126063 | AK007838.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126063 | AK008265.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 305364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288536 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318450 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337387 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3550 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88172 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123129 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126063 | AK003602.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126063 | NM_078479.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126063 | AK007662.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126063 | AK007838.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126063 | AK008265.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509012 | AK003462.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509012 | AK003602.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509012 | NM_078479.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509012 | AB049956.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509012 | AK002588.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346100 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123129 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00703285 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00703288 | - | - |
| Whole Mouse Genome G4122A | A_51_P509012 | AK003462.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509012 | AK003602.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509012 | NM_078479.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509012 | AB049956.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509012 | AK002588.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118456 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121918 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102118 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123129 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00703288 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02438392 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02628063 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02438386 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02878927 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02438391 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02438384 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02878925 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02438390 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346101 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346100 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346103 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346102 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02809728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02873868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02572552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02873865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02878927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02878925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02283537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03132694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02539844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03106562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02886875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02438391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02942585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02991830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02327519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02628063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02942586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02539843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02333348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03030389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02438387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02873866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02327101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02991832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02306225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02991829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03152845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02539853 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10500269 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500271 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500270 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422451_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10500268 | - | - |
| Mouse Genome 430 2.0 Array | 1422451_at | - | - |
| Mouse Genome 430 2.0 Array | 1422451_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459677_at | - | - |
| Mouse Genome 430 2.0 Array | 1459677_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66292_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66292_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300019376.mm.1 | NM_078479.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019376.mm.1 | NM_078479.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019376.mm.1 | TC0300002488.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019380.mm.1 | NM_078479.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019380.mm.1 | NM_078479.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019380.mm.1 | TC0300002488.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019377.mm.1 | NM_078479.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019377.mm.1 | NM_078479.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019377.mm.1 | TC0300002488.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019375.mm.1 | NM_078479.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019375.mm.1 | NM_078479.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019375.mm.1 | TC0300002488.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422451_at | - | - |
| Mouse Expression Set 430 | 1422451_PM_at | - | - |
| Murine Genome U74v2 Set | 94912_PM_at | - | - |
| Murine Genome U74v2 Set | 94912_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422451_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422451_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422451_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422451_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2650008 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002650008 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2745555 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2745555 | - | - |
| mouse-6 v1.1 (Illumina) | scl066292.2_43-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2650008 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066292.2_43-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650008 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2745555 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2745555 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066292.2_43-S | - | - |

Mrps21
Genomic Map