Assay Details
| Gene Symbol: Tnfrsf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK048111.1 | - | 1 | 1717 | 79 | |
| AK081533.1 | - | 1 | 1717 | 79 | ||
| BC076592.1 | - | 6 | 4098 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tnfrsf22
Genomic Map