Assay Details
| Gene Symbol: Ifna14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206975.1 | NP_996858.1 | 1 | 360 | 161 | |
| GenBank mRNA | BC120722.1 | - | 1 | 410 | 40129645 | 161 |
| BC125321.1 | - | 1 | 410 | 40129836 | 161 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
humoral immune response
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
cytokine activity
type I interferon receptor binding
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
humoral immune response
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
cytokine activity
type I interferon receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996365 | NM_206975.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996365 | BC120722.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996365 | BC125321.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996365 | NM_206975.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996365 | BC120722.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996365 | BC125321.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091237 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140446 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03275383 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03275392 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03275381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03224001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03421293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03982329 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03230539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091253 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10514293 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10514294 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10514295 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10514292 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400024298.mm.1 | TC0400003003.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400024299.mm.1 | TC0400003003.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400024299.mm.1 | NM_206975.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400024299.mm.1 | NM_206975.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400024300.mm.1 | TC0400003003.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422404_x_at | - | - |
| Mouse Expression Set 430 | 1422403_PM_at | - | - |
| Mouse Expression Set 430 | 1422403_at | - | - |
| Mouse Expression Set 430 | 1422404_PM_x_at | - | - |
| Murine Genome U74v2 Set | 102149_f_at | - | - |
| Murine Genome U74v2 Set | 102149_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422404_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422404_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422403_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422403_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 428429 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103610577 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103610577 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4540142 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004540142 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2617487 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2687005 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2617487 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226397 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2687005 | - | - |
| mouse-6 v1.1 (Illumina) | scl0404549.1_330-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_46195447-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4540142 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103610577 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0404549.1_330-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540142 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103610577 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2687005 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226397 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2687005 | - | - |

Ifna14
Genomic Map