Assay Details
Gene Symbol: Ywhah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011738.2 | NP_035868.1 | 1 | 107 | ||
GenBank mRNA | AF077002.1 | - | 1 | 107 | ||
AK077596.1 | - | 1 | 107 | |||
AK083936.1 | - | 1 | 107 | |||
AK149224.1 | - | 1 | 107 | |||
AK168520.1 | - | 1 | 107 | |||
AK169035.1 | - | 1 | 107 | |||
BC061497.1 | - | 1 | 4949445 | 107 | ||
CB183421.1 | - | 1 | 107 | |||
D87661.1 | - | 1 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF077002.1, AK077596.1, AK083936.1, AK149224.1, AK168520.1, AK169035.1, BC061497.1, D87661.1, NM_011738.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
