Assay Details
| Gene Symbol: Flg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF510860.1 | - | 1 - 2 | 142 | 89 | |
| AI119275.1 | - | 2 - 3 | 190 | 89 | ||
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170289 | J03458.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170289 | AK076261.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170289 | AV230893.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170289 | AW107830.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915966 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170289 | J03458.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170289 | AK076261.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170289 | AV230893.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170289 | AW107830.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P127877 | AF510860.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P127877 | AF510860.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050415 | J03458.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200186 | J03458.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101522 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03103038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02488845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02454515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335378 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493926 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493928 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493927 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493931 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493932 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427268_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493925 | - | - |
| Mouse Genome 430 2.0 Array | 1427268_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427268_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14246_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14246_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa16520_at | - | - |
| Mouse Expression Set 430 | 1427268_at | - | - |
| Mouse Expression Set 430 | 1427268_PM_at | - | - |
| Mouse Expression Set 430 | 1451613_at | - | - |
| Mouse Expression Set 430 | 1451613_PM_at | - | - |
| Murine Genome U74v2 Set | 100420_at | - | - |
| Murine Genome U74v2 Set | 161981_PM_r_at | - | - |
| Murine Genome U74v2 Set | 161981_r_at | - | - |
| Murine Genome U74v2 Set | 100420_PM_at | - | - |
| Murine Genome U74v2 Set | 166991_PM_at | - | - |
| Murine Genome U74v2 Set | 166991_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451613_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427268_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451613_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427268_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427268_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427268_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 569022 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104760609 | - | - |
| MouseWG-6 v2 Expression BeadChip | 103450037 | - | - |

Flg
Genomic Map