Assay Details
| Gene Symbol: Hotair | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_047528.1 | 1 - 2 | 444 | 86 | ||
| GenBank mRNA | AK035706.1 | - | 1 - 2 | 228 | 86 | |
| BB628325.1 | - | 1 - 2 | 225 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hotair
Genomic Map