Assay Details
| Gene Symbol: 2310068J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028124.1 | 1 | 950 | 139 | ||
| GenBank mRNA | AK010113.1 | - | 1 | 952 | 139 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
La ribonucleoprotein domain family, member 4 |
Larp4 |
DXErtd793, DXErtd793e |
Anti-sense | Chr. 15 - 99970065 - 100016358 on NCBI Build GRCm38 | |
| Downstream | DIP2 disco-interacting protein 2 homolog B (Drosophila) | Dip2b |
4932422C22, AI317237, AI854602, mKIAA1463 |
65.0kb | Anti-sense | Chr. 15 - 100038318 - 100219474 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2310068J16Rik
Genomic Map