Assay Details
| Gene Symbol: Ntng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011411.1 | - | 1 | 1470 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | senataxin | Setx |
A130090N03, A930037J23Rik, AOA2, AW060766, Als4, SCAR1, Sen1, mKIAA0625 |
-30.3kb | Anti-sense | Chr. 2 - 29122588 - 29182471 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 6530402F18 gene | 6530402F18Rik | mKIAA1857 | 30.8kb | Sense | Chr. 2 - 29245116 - 29252993 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ntng2
Genomic Map