Assay Details
| Gene Symbol: Zfp81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207541.1 | NP_997424.1 | 4 | 1925 | 104 | |
| GenBank mRNA | AK079439.1 | - | 2 | 1084 | 104 | |
| AK135853.1 | - | 4 | 1926 | 104 | ||
| BB143786.2 | - | 1 | 542 | 104 | ||
| BC026640.1 | - | 1 | 1348 | 104 | ||
| BC117831.1 | - | 4 | 1875 | 40092935 | 104 | |
| BC117832.1 | - | 4 | 1877 | 40092936 | 104 | |
More Information
Additional Information:
				    	 
					    	
					        	
					            	Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			Set Membership: | 
		   
					
						
							 | 
				
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
					
						
							Agilent
					
						
							Affymetrix
					
						
							ABI
					
						
							Illumina
					
					
					
						
							
								
						
					
						
							
								
						
					
						
							
								
						
					
						
							
								
						
					
				
			| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31148787 | - | - | 
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31148785 | - | - | 
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31148784 | - | - | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046478 | BC026640.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046478 | BC117831.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046478 | BC117832.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046478 | AK135853.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046478 | NM_207541.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277629 | BC026640.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277629 | BB143786.2 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277629 | AK135853.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277629 | NM_207541.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277629 | AK079439.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277629 | BC026640.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277629 | BB143786.2 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277629 | AK135853.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277629 | NM_207541.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P277629 | AK079439.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046485 | AK135853.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046485 | NM_207541.1 | mouse_transcriptome | 
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935507 | - | - | 
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525807 | - | - | 
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545768 | - | - | 
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77742 | - | - | 
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48687 | - | - | 
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484123 | - | - | 
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 567303 | - | - | 
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105190 | - | - | 
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 90225 | - | - | 
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 122319 | - | - | 
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116678 | - | - | 
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108250 | - | - | 
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133605 | - | - | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046478 | BC026640.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046478 | BC117831.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046478 | BC117832.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046478 | AK135853.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046478 | NM_207541.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277629 | BC026640.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277629 | BB143786.2 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277629 | AK135853.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277629 | NM_207541.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277629 | AK079439.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277629 | BC026640.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277629 | BB143786.2 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277629 | AK135853.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277629 | NM_207541.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P277629 | AK079439.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046485 | AK135853.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046485 | NM_207541.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P277629 | BC026640.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P277629 | BB143786.2 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P277629 | AK135853.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P277629 | NM_207541.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P277629 | AK079439.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P277629 | BC026640.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P277629 | BB143786.2 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P277629 | AK135853.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P277629 | NM_207541.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P277629 | AK079439.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_52_P510038 | BC026640.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_52_P510038 | BC117831.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_52_P510038 | BC117832.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_52_P510038 | BB143786.2 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_52_P510038 | AK135853.1 | mouse_transcriptome | 
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31148784 | - | - | 
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31148787 | - | - | 
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31148785 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148733 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148752 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148707 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148744 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148757 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148787 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148710 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148769 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148724 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148717 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148754 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148796 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148768 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148701 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148702 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148730 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148789 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148743 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148747 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148798 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148792 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148748 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148712 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148784 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148718 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148700 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148704 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148782 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148779 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148708 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148735 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148749 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148741 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148791 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148722 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148759 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148775 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148739 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148764 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148788 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148728 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148716 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148697 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13129618 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13281516 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148782 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148794 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148789 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148781 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148784 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148787 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148776 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13364872 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148779 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148791 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13158937 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148796 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148775 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148773 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148780 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13129619 | - | - | 
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148798 | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10449994 | - | - | 
| Affymetrix Mouse Gene 1.0 ST Array | 10449998 | - | - | 
| Affymetrix Mouse Gene 1.0 ST Array | 10449993 | - | - | 
| Affymetrix Mouse Gene 1.0 ST Array | 10449992 | - | - | 
| Affymetrix Mouse Gene 1.0 ST Array | 10449995 | - | - | 
| Affymetrix Mouse Gene 1.0 ST Array | 10449997 | - | - | 
| Affymetrix Mouse Gene 1.0 ST Array | 10449996 | - | - | 
| Affymetrix HT MG-430 PM Array Plate | 1444076_PM_at | - | - | 
| Affymetrix HT MG-430 PM Array Plate | 1457685_PM_at | - | - | 
| Affymetrix HT MG-430 PM Array Plate | 1427499_PM_at | - | - | 
| Affymetrix Mouse Gene 1.1 ST Array | 10449991 | - | - | 
| Mouse Genome 430 2.0 Array | 1457685_at | - | - | 
| Mouse Genome 430 2.0 Array | 1457685_PM_at | - | - | 
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224694_PM_at | - | - | 
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224694_at | - | - | 
| Affymetrix Mouse Clariom S Array | PSR1700017271.mm.1 | NM_207541.1.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR1700017271.mm.1 | NM_207541.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR1700017271.mm.1 | TC1700001869.mm.1 | MTA TC | 
| Affymetrix HT Mouse Genome 430B Array | 1457685_at | - | - | 
| Affymetrix HT Mouse Genome 430B Array | 1457685_PM_at | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 393391 | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 106980092 | - | - | 
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2624853 | - | - | 
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2624853 | - | - | 
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226845 | - | - | 
| Illumina Sentrix mouse-6.v1 bead array | 106980092 | - | - | 
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106980092 | - | - | 
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226845 | - | - | 

Zfp81
Genomic Map