Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100007486.mm.1 |
NM_011496.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007486.mm.1 |
TC1100000892.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007491.mm.1 |
NM_011496.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007491.mm.1 |
TC1100000892.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007494.mm.1 |
NM_011496.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007494.mm.1 |
TC1100000892.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007493.mm.1 |
NM_011496.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007493.mm.1 |
TC1100000892.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007488.mm.1 |
NM_011496.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007488.mm.1 |
TC1100000892.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007495.mm.1 |
NM_011496.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007495.mm.1 |
TC1100000892.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007485.mm.1 |
NM_011496.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007485.mm.1 |
TC1100000892.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007484.mm.1 |
NM_011496.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007484.mm.1 |
TC1100000892.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007498.mm.1 |
NM_011496.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007498.mm.1 |
TC1100000892.mm.1 |
MTA TC |