Assay Details
| Gene Symbol: 4933406I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029438.1 | 2 - 3 | 521 | 119 | ||
| GenBank mRNA | AK016630.1 | - | 2 - 3 | 382 | 119 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
proteasome (prosome, macropain) subunit, alpha type 1 |
Psma1 |
C2, HC2, Pros-30, alpha-type |
-84.4kb | Sense | Chr. 7 - 114264550 - 114276116 on NCBI Build GRCm38 |
| Downstream |
phosphodiesterase 3B, cGMP-inhibited |
Pde3b |
9830102A01Rik, AI847709 |
18.7kb | Anti-sense | Chr. 7 - 114414700 - 114537938 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4933406I18Rik
Genomic Map