Assay Details
| Gene Symbol: Cramp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020608.3 | NP_065633.2 | 15 - 16 | 3014 | 66 | |
| GenBank mRNA | AK089393.1 | - | 15 - 16 | 3274 | 66 | |
| AK129356.1 | - | 6 - 7 | 1190 | 66 | ||
| AK147411.1 | - | 15 - 16 | 3015 | 66 | ||
| AK147449.1 | - | 15 - 16 | 3015 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cramp1l
Genomic Map