Assay Details
| Gene Symbol: Cramp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020608.3 | NP_065633.2 | 18 - 19 | 3486 | 88 | |
| GenBank mRNA | AK129356.1 | - | 9 - 10 | 1662 | 88 | |
| AK147411.1 | - | 18 - 19 | 3487 | 88 | ||
| AK147449.1 | - | 18 - 19 | 3487 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cramp1l
Genomic Map