Assay Details
| Gene Symbol: Gm32849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045483.1 | - | 1 - 2 | 108 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC545966 | LOC545966 | -172.1kb | Sense | Chr. 7 - 48877958 - 48913067 on NCBI Build GRCm38 | |
| Downstream | developing brain homeobox 1 | Dbx1 |
AI426026, Dbx |
545.6kb | Anti-sense | Chr. 7 - 49631499 - 49636835 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm32849
Genomic Map