Assay Details
Gene Symbol: C330013F16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045456.1 | 3 - 4 | 317 | 158 | ||
GenBank mRNA | AK021198.1 | - | 3 - 4 | 316 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P131324 | BB283810.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P131324 | AK021198.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92874 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780008 | BC030305.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593573 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 565341 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 887885 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 71723 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603544 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275257 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608138 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 429638 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 85257 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137286 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 796530 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 785823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510361 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229222 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632705 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42904 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336205 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 594683 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569462 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597298 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827714 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632786 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819676 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 87098 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 496215 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215729 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205313 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 242245 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624380 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 169864 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204178 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771243 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 231383 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 403591 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927300 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176814 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198211 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344219 | AK007350.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344219 | AK146311.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344219 | AK152351.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344219 | AK171926.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344219 | BC021887.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344219 | BC037140.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344219 | BC055896.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344219 | BC062200.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344219 | NM_183308.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644686 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365667 | AF268066.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365667 | AF281278.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365667 | BC119182.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365667 | BC119184.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365667 | NM_021320.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704588 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 94188 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 799429 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320699 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573700 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351176 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 849800 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 177540 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405877 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 146627 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858878 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318795 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45462 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30178 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46581 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8127 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79822 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129477 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121077 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 95601 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 81255 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156350 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P131324 | BB283810.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P131324 | AK021198.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435559 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435752 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435708 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435761 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03763666 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435668 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156350 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435608 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03763769 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435578 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435543 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03763705 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435781 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09435741 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131324 | BB283810.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131324 | AK021198.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09435570 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156350 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09435713 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03763769 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03763666 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693902 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693666 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693854 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15003914 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693861 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693705 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693672 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693859 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693914 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693710 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693569 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14677241 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693860 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693896 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693878 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14728501 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693867 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693871 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693852 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14556355 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15003925 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693566 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693888 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693856 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693885 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693663 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693910 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693673 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693570 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14582455 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693668 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693851 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693573 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14656454 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14582454 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693911 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14716776 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693564 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693893 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693567 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693892 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693857 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693903 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693881 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15097475 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693907 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693883 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15065120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693704 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693659 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14534787 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693689 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693576 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693706 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693572 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693909 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693901 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693562 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693862 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693872 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693671 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693866 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693688 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14886247 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693684 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15186319 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693880 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693869 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693886 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693658 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14656458 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693916 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693894 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693703 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693662 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693708 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693849 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693863 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693890 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693574 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693670 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32693685 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0X00019183.mm.1 | NR_045456.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00019183.mm.1 | NR_045456.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00019183.mm.1 | TC0X00003059.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 495958 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 106860390 | - | - |
