Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700022696.mm.1 |
TC0700002627.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022696.mm.1 |
NM_001031667.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022696.mm.1 |
NM_001031667.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022697.mm.1 |
TC0700002627.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022697.mm.1 |
NM_001031667.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022697.mm.1 |
NM_001031667.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022698.mm.1 |
TC0700002627.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022698.mm.1 |
NM_001031667.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022698.mm.1 |
NM_001031667.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022699.mm.1 |
TC0700002627.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022699.mm.1 |
NM_001031667.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022699.mm.1 |
NM_001031667.1 |
Refseq |