Assay Details
Gene Symbol: Gm7607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK027968.1 | - | 1 - 2 | 183 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P875976 | AK027968.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P875976 | AK027968.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P875976 | AK027968.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P875976 | AK027968.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140091 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07478113 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318538 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318579 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07478131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07794858 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318525 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318574 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318500 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318518 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318559 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08023581 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318555 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318517 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318578 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08130137 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318584 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07554864 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318581 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318562 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07816449 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318561 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318528 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318498 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318508 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318531 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318564 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318507 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318526 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318534 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318520 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318529 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08189127 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07554868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07478130 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318506 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318541 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07657195 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318540 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318536 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318566 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318522 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318516 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07478117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26318515 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08158780 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07794861 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 437939 | - | - |
