Assay Details
| Gene Symbol: 4930562D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016189.1 | - | 1 - 2 | 77 | 150 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 10635 | Gm10635 | ENSMUSG00000074174 | -169.5kb | Anti-sense | Chr. 9 - 79444037 - 79519302 on NCBI Build GRCm38 |
| Downstream | cytochrome c oxidase subunit VIIa 2 | Cox7a2 |
COX7AL, Cox7a3, CoxVIIa-L |
64.3kb | Anti-sense | Chr. 9 - 79755241 - 79759853 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930562D21Rik
Genomic Map