| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400026996.mm.1 |
TC0400003368.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026996.mm.1 |
NM_001145637.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026996.mm.1 |
NM_001145637.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026993.mm.1 |
TC0400003368.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026993.mm.1 |
NM_001145637.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026993.mm.1 |
NM_001145637.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026994.mm.1 |
TC0400003368.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400026994.mm.1 |
NM_001145637.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400026994.mm.1 |
NM_001145637.1 |
Refseq |
| Murine Genome U74v2 Set |
171086_PM_at |
- |
- |
| Murine Genome U74v2 Set |
171086_at |
- |
- |