Assay Details
Gene Symbol: Gm40038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK052977.1 | - | 1 - 2 | 198 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | progesterone receptor membrane component 2 | Pgrmc2 |
4631434O19Rik, 5730409G06Rik, DG6, PMBP |
-80.0kb | Sense | Chr. 3 - 41066326 - 41083046 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 1700052H01 gene | 1700052H01Rik | 180.8kb | Anti-sense | Chr. 3 - 41353419 - 41404456 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
