Assay Details
| Gene Symbol: Gm38414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082764.1 | - | 1 - 2 | 194 | 164 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DNA segment, Chr 8, ERATO Doi 82, expressed |
D8Ertd82e |
9830148H23, NACK, Sgk223, mFLJ00269 |
-8.2kb | Sense | Chr. 8 - 36094625 - 36147787 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 39164 |
Gm39164 | 17.7kb | Anti-sense | Chr. 8 - 36174140 - 36176075 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm38414
Genomic Map