Assay Details
Gene Symbol: LOC432658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK048895.1 | - | 1 - 2 | 66 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | A kinase (PRKA) anchor protein 6 | Akap6 |
AI482140, Akapalpha, Akapbeta, PRKA |
Anti-sense | Chr. 12 - 52699016 - 53155568 on NCBI Build GRCm38 | |
Downstream | neuronal PAS domain protein 3 | Npas3 |
4930423H22Rik, bHLHe12 |
490.0kb | Anti-sense | Chr. 12 - 53248003 - 54072175 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
