Assay Details
| Gene Symbol: Gm26629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039031.1 | - | 1 - 2 | 111 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 30123 |
Gm30123 | -344.0kb | Sense | Chr. 19 - 50324030 - 50333430 on NCBI Build GRCm38 | |
| Downstream | insulin I | Ins1 |
Ins-1, Ins2-rs1 |
1.5mb | Sense | Chr. 19 - 52264323 - 52265476 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm26629
Genomic Map