Assay Details
| Gene Symbol: Rbm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167776.1 | NP_001161248.1 | 1 - 2 | 296 | 83 | |
| NM_145627.2 | NP_663602.1 | 1 - 2 | 296 | 83 | ||
| GenBank mRNA | AK041871.1 | - | 1 - 2 | 271 | 83 | |
| AK089105.1 | - | 1 - 2 | 107 | 83 | ||
| AK122208.1 | - | 1 - 2 | 214 | 83 | ||
| AK145991.1 | - | 1 - 2 | 296 | 83 | ||
| AK167666.1 | - | 1 - 2 | 274 | 83 | ||
| BC093519.1 | - | 1 - 2 | 228 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041871.1,AK089105.1,AK145991.1,AK167666.1,BC093519.1,NM_001167776.1,NM_145627.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbm10
Genomic Map