Assay Details
Gene Symbol: Tarbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253795.1 | NP_001240724.1 | 1 - 2 | 410 | 66 | |
GenBank mRNA | AK144205.1 | - | 1 - 2 | 248 | 66 | |
BC053432.1 | - | 1 - 2 | 164 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
