Assay Details
| Gene Symbol: Ywhaz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253805.1 | NP_001240734.1 | 1 - 2 | 395 | 70 | |
| GenBank mRNA | AK083368.1 | - | 1 - 2 | 123 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK083368.1, NM_001253805.1 transcripts.
Set Membership: |
|

Ywhaz
Genomic Map