Assay Details
Gene Symbol: Med29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026042.3 | NP_080318.2 | 2 - 3 | 296 | 97 | |
GenBank mRNA | AK005112.1 | - | 2 - 3 | 298 | 97 | |
AK013006.1 | - | 2 - 3 | 296 | 97 | ||
BC023192.1 | - | 2 - 3 | 279 | 4947722 | 97 | |
BC083314.1 | - | 2 - 3 | 278 | 6823222 | 97 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
