Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400016674.mm.1 |
NM_001164327.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016674.mm.1 |
NM_001164327.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016674.mm.1 |
TC1400002162.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016676.mm.1 |
NM_001164327.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016676.mm.1 |
NM_001164327.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016676.mm.1 |
TC1400002162.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016678.mm.1 |
NM_001164327.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016678.mm.1 |
NM_001164327.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016678.mm.1 |
TC1400002162.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016679.mm.1 |
NM_001164327.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016679.mm.1 |
NM_001164327.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016679.mm.1 |
TC1400002162.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016681.mm.1 |
NM_001164327.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016681.mm.1 |
NM_001164327.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016681.mm.1 |
TC1400002162.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016680.mm.1 |
NM_001164327.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016680.mm.1 |
NM_001164327.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016680.mm.1 |
TC1400002162.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016673.mm.1 |
NM_001164327.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016673.mm.1 |
NM_001164327.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016673.mm.1 |
TC1400002162.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016675.mm.1 |
NM_001164327.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016675.mm.1 |
NM_001164327.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016675.mm.1 |
TC1400002162.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016677.mm.1 |
NM_001164327.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016677.mm.1 |
NM_001164327.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016677.mm.1 |
TC1400002162.mm.1 |
MTA TC |