Assay Details
| Gene Symbol: Zfp330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145600.1 | NP_663575.1 | 1 - 2 | 88 | ||
| GenBank mRNA | AK050534.1 | - | 1 - 2 | 88 | ||
| AK086617.1 | - | 1 - 2 | 88 | |||
| BC008086.1 | - | 1 - 2 | 3586161 | 88 | ||
| BC030675.1 | - | 1 - 2 | 71 | 1068780 | 88 | |
| BC083081.1 | - | 1 - 2 | 107 | 6510396 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050534.1, AK086617.1, BC008086.1, BC030675.1, BC083081.1, NM_145600.1 transcripts.
Set Membership: |
|

Zfp330
Genomic Map