Assay Details
| Gene Symbol: Ccnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183015.3 | NP_898836.2 | 12 - 13 | 4105 | 84 | |
| GenBank mRNA | AJ416459.1 | - | 4 - 5 | 502 | 84 | |
| AJ555464.1 | - | 12 - 13 | 4106 | 84 | ||
| AJ555465.1 | - | 11 - 12 | 1381 | 84 | ||
| BC132411.1 | - | 11 - 12 | 4059 | 40130688 | 84 | |
| BC138355.1 | - | 11 - 12 | 4059 | 8861375 | 84 | |
More Information
Set Membership: |
|

Ccnb3
Genomic Map