Assay Details
| Gene Symbol: Zfp758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028301.1 | - | 1 - 2 | 237 | 140 | |
| BC056632.1 | - | 1 - 2 | 5036441 | 140 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028301.1 transcripts.
Set Membership: |
|

Zfp758
Genomic Map