| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10599288 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599290 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599247 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599287 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599250 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10599248 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10599286 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10599246 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0X00002383.mm.1 |
NM_001081669.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002383.mm.1 |
NM_001081669.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002383.mm.1 |
TC0X00000347.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002384.mm.1 |
NM_001081669.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002384.mm.1 |
NM_001081669.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002384.mm.1 |
TC0X00000347.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002385.mm.1 |
NM_001081669.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002385.mm.1 |
NM_001081669.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002385.mm.1 |
TC0X00000347.mm.1 |
MTA TC |