Assay Details
| Gene Symbol: Actg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009609.2 | NP_033739.1 | 6 | 1850 | 154 | |
| GenBank mRNA | AK012251.1 | - | 6 | 1862 | 154 | |
| AK076081.1 | - | 6 | 1859 | 154 | ||
| AK152202.1 | - | 6 | 1863 | 154 | ||
| AK153446.1 | - | 6 | 1861 | 154 | ||
| AK157726.1 | - | 6 | 1859 | 154 | ||
| AK159867.1 | - | 6 | 1862 | 154 | ||
| BC003337.1 | - | 6 | 1845 | 3257312 | 154 | |
| BC008258.1 | - | 1 | 672 | 154 | ||
| BC018571.1 | - | 6 | 1845 | 154 | ||
| BC021796.1 | - | 6 | 1847 | 4218945 | 154 | |
| BC023248.1 | - | 6 | 1850 | 3499390 | 154 | |
| BC099371.1 | - | 6 | 1842 | 30841085 | 154 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183970 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183963 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183957 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183973 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183966 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183969 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183960 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183968 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183965 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183958 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183977 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183961 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183974 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183959 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183962 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183972 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P72237 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985984 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008907 | X13055.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008907 | BC023248.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008907 | BC099371.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008907 | M21495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008907 | BC003337.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114993 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1970597 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142172 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820864 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P72237 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008907 | X13055.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008907 | BC023248.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008907 | BC099371.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008907 | M21495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008907 | BC003337.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114993 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985984 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1970597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28183960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28183953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28183957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28183958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28183947 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114990 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136570 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135151 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115217 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09098336 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09098339 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09098343 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183961 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183974 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183977 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183972 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183957 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183968 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183973 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183969 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183965 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183966 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183970 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183963 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183960 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183959 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183962 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28183958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28183963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03208536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03422584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03422585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03218172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03208535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03218177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03906138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03218176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23166898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09743763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09638689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09651386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09098343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09743762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09032818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09638695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09394550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09394580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09394561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183931 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10393759 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393757 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393755 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393756 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571654 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393760 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393758 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406418 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415779_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10571653 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10506424 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10393754 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10406417 | - | - |
| Mouse Genome 430 2.0 Array | 1415779_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1415779_PM_s_at | - | - |
| Mouse Expression Set 430 | 1415779_PM_s_at | - | - |
| Mouse Expression Set 430 | 1415779_s_at | - | - |
| Murine Genome U74v2 Set | 96573_at | - | - |
| Murine Genome U74v2 Set | 96573_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415779_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415779_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415779_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415779_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 540215 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 540215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000540215 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2674763 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2674763 | - | - |
| mouse-6 v1.1 (Illumina) | scl011465.1_245-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 540215 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl011465.1_245-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 540215 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2674763 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2674763 | - | - |

Actg1
Genomic Map