Assay Details
| Gene Symbol: Pisd-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003517.1 | 7 | 126 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
Sfi1 homolog, spindle assembly associated (yeast) |
Sfi1 | Anti-sense | Chr. 11 - 3131850 - 3203343 on NCBI Build GRCm38 | ||
| Downstream | eukaryotic translation initiation factor 4E nuclear import factor 1 | Eif4enif1 |
2610509L04Rik, A930019J01Rik, AA410001, AU021239, Clast4, D11Ertd166e |
69.7kb | Sense | Chr. 11 - 3201598 - 3244588 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Pisd-ps1
Genomic Map