Assay Details
| Gene Symbol: Akap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019921.3 | NP_064305.2 | 6 - 7 | 1241 | 67 | |
| GenBank mRNA | AK049399.1 | - | 6 - 7 | 1210 | 67 | |
| AK051044.1 | - | 4 - 5 | 876 | 67 | ||
| AK083255.1 | - | 6 - 7 | 1210 | 67 | ||
| BC038483.1 | - | 3 - 4 | 299 | 67 | ||
| BC054105.1 | - | 3 - 4 | 544 | 30022728 | 67 | |
More Information
Set Membership: |
|

Akap10
Genomic Map