Assay Details
| Gene Symbol: Apol10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177820.4 | NP_808488.2 | 4 | 845 | 106 | |
| GenBank mRNA | AK033678.1 | - | 817 | 106 | ||
| AK144113.1 | - | 4 | 849 | 106 | ||
| BC147348.1 | - | 791 | 9056388 | 106 | ||
| BC147349.1 | - | 791 | 9056408 | 106 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006494 | AK033678.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006494 | NM_177820.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P120380 | BB653856.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P120380 | BB634068.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P120380 | BC147349.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P120380 | NM_001143686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P120380 | AK039744.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 449434 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819261 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823556 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 165509 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740560 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588598 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916244 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16671 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153906 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160977 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006494 | AK033678.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006494 | NM_177820.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P120380 | BB653856.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P120380 | BB634068.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P120380 | BC147349.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P120380 | NM_001143686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P120380 | AK039744.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160977 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03075682 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153906 | - | - |
| Whole Mouse Genome G4122A | A_51_P410387 | BC147349.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P410387 | AK033678.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P410387 | BC147348.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P410387 | NM_177820.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P410387 | AK033521.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135956 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160977 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153906 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11938832 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11866333 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11866336 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11866344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30354785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11938832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12058557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12007422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12058555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12098624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11892437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11866344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30354785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12401143 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10430192 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430193 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430191 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430190 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 573789 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 780731 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 780731 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000780731 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218964 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218964 | - | - |
| mouse-6 v1.1 (Illumina) | scl47012.4_666-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 780731 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47012.4_666-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 780731 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2756223 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2756223 | - | - |

Apol10b
Genomic Map