Assay Details
Gene Symbol: Trh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009426.3 | NP_033452.2 | 3 | 1268 | 122 | |
GenBank mRNA | AI836004.1 | - | 1 | 122 | ||
AK010666.1 | - | 3 | 1266 | 122 | ||
BC053493.1 | - | 3 | 1251 | 30010395 | 122 | |
X59387.1 | - | 2 | 1221 | 122 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
histamine metabolic process
adult walking behavior
hormone-mediated signaling pathway
negative regulation of glutamate secretion
positive regulation of gamma-aminobutyric acid secretion
positive regulation of insulin secretion
eating behavior
negative regulation of feeding behavior
hormone activity
neuropeptide hormone activity
protein binding
thyrotropin-releasing hormone activity
histamine metabolic process
adult walking behavior
hormone-mediated signaling pathway
negative regulation of glutamate secretion
positive regulation of gamma-aminobutyric acid secretion
positive regulation of insulin secretion
eating behavior
negative regulation of feeding behavior
hormone activity
neuropeptide hormone activity
protein binding
thyrotropin-releasing hormone activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071349 | NM_009426.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071349 | AK010666.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071349 | BB400231.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071349 | BC053493.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30582 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45076 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150295 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071349 | NM_009426.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071349 | AK010666.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071349 | BB400231.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071349 | BC053493.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338278 | X59387.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338278 | NM_009426.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338278 | AK010666.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338278 | BC053493.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644002 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644005 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644027 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644023 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24643995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644004 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644019 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644032 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644010 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644034 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644014 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644036 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644040 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24643991 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644041 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24643990 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24643999 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24643997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644038 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24643996 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644012 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644008 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644016 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644024 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24643998 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644020 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644029 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644017 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644021 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644037 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644025 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644007 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24644033 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150295 | - | - |
Whole Mouse Genome G4122A | A_51_P338278 | X59387.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P338278 | NM_009426.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P338278 | AK010666.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P338278 | BC053493.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108566 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150295 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05238465 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05238474 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05120061 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05120058 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05238466 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05238472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643963 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24643986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24644019 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05224381 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05224379 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643995 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644008 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05307942 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05623990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05497554 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05560207 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05238465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644054 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05348812 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644040 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644033 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643960 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643963 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05048339 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05430930 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05502670 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643968 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05396620 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05502672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644060 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05592157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643965 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05238464 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05348797 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05556375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643985 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05048340 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643967 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644027 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05623989 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05556377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643966 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05238474 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643961 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05348796 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05348795 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644045 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644046 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05238472 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644002 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05120061 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644051 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24643996 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05120058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644009 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05302003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05348798 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24644001 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10546419 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10546420 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10546418 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418756_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10546417 | - | - |
Mouse Genome 430 2.0 Array | 1418756_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418756_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22044_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22044_PM_at | - | - |
Mouse Expression Set 430 | 1418756_PM_at | - | - |
Mouse Expression Set 430 | 1418756_at | - | - |
Murine Genome U74v2 Set | 102665_at | - | - |
Murine Genome U74v2 Set | 102665_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418756_at | - | - |
Mouse Genome 430A 2.0 Array | 1418756_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418756_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418756_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 759955 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5050576 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5050576 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005050576 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237934 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1237934 | - | - |
mouse-6 v1.1 (Illumina) | scl28676.3_567-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5050576 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl28676.3_567-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5050576 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1237934 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237934 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl28676.3_567-S | - | - |
