Assay Details
Gene Symbol: Vmn1r51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011683.2 | NP_035813.2 | 3 | 1905 | 151 | |
GenBank mRNA | BC046308.1 | - | 3 | 1905 | 6306893 | 151 |
Y12725.1 | - | 2 | 1875 | 151 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055697 | Y12725.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055697 | BY430696.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055697 | NM_011683.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36606 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635179 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549668 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382110 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187240 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121074 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153455 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055697 | Y12725.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055697 | BY430696.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055697 | NM_011683.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P512439 | Y12725.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153455 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05977710 | - | - |
Whole Mouse Genome G4122A | A_51_P512439 | Y12725.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100962 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125585 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108189 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630963 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24631001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630959 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630919 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630929 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05570985 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05717768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630937 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630961 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630963 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05571001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05717761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05717772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05861439 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05717770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630975 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630970 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630943 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05717778 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630957 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630939 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05844876 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630950 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05570984 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630928 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05844872 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05861431 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630955 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630927 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630953 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630914 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630925 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24630959 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10539970 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10539972 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10539971 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450558_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10539969 | - | - |
Mouse Genome 430 2.0 Array | 1450558_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1450558_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22296_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22296_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600008260.mm.1 | TC0600001062.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600008260.mm.1 | NM_011683.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600008260.mm.1 | NM_011683.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600008256.mm.1 | TC0600001062.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600008256.mm.1 | NM_011683.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600008256.mm.1 | NM_011683.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600008257.mm.1 | TC0600001062.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600008257.mm.1 | NM_011683.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600008257.mm.1 | NM_011683.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600008259.mm.1 | TC0600001062.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600008259.mm.1 | NM_011683.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600008259.mm.1 | NM_011683.2 | Refseq |
Mouse Expression Set 430 | 1450558_PM_at | - | - |
Mouse Expression Set 430 | 1450558_at | - | - |
Murine Genome U74v2 Set | 94571_at | - | - |
Murine Genome U74v2 Set | 94571_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450558_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450558_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450558_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450558_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 601430 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2190301 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2190301 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002190301 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2455370 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2455370 | - | - |
mouse-6 v1.1 (Illumina) | scl29760.3.1_197-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2190301 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl29760.3.1_197-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2190301 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2931315 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2455370 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2931315 | - | - |
