Assay Details
| Gene Symbol: Tomm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099675.1 | NP_001093145.1 | 2 | 234 | 115 | |
| NM_001134646.1 | NP_001128118.1 | 3 | 284 | 165 | ||
| GenBank mRNA | AA423234.1 | - | 2 | 234 | 115 | |
| AK003822.1 | - | 2 | 230 | 115 | ||
| CA466100.1 | - | 3 | 276 | 165 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063598 | CA466100.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063598 | NM_001099675.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063598 | NM_001134646.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063598 | AK003822.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063598 | AW047671.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062811 | CA466100.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062811 | NM_001099675.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062811 | NM_001134646.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062811 | AK003822.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062811 | AW047671.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392696 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86303 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41010 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152285 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062811 | CA466100.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062811 | NM_001099675.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062811 | NM_001134646.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062811 | AK003822.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062811 | AW047671.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063598 | CA466100.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063598 | NM_001099675.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063598 | NM_001134646.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063598 | AK003822.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063598 | AW047671.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152285 | - | - |
| Whole Mouse Genome G4122A | A_52_P40212 | CA466100.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P40212 | NM_001099675.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P40212 | NM_001134646.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P40212 | AA423234.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P40212 | AK003822.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121613 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22881157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03818156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03310520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04004773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03631069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03460790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04004780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03286973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03936366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03436241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04004781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03744216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04083824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03995217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03460787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03224035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03779588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03936369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03436237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22881159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03436243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03744218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03310518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03779586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03294790 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10512702 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512703 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415733_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10512701 | - | - |
| Mouse Genome 430 2.0 Array | 1415733_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1415733_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68512_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68512_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400020983.mm.1 | TC0400002572.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020983.mm.1 | NM_001099675.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020983.mm.1 | NM_001099675.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020983.mm.1 | NM_001134646.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020983.mm.1 | NM_001134646.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020986.mm.1 | TC0400002572.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400020986.mm.1 | NM_001134646.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400020986.mm.1 | NM_001134646.1 | Refseq |
| Mouse Expression Set 430 | 1415733_PM_a_at | - | - |
| Mouse Expression Set 430 | 1415733_a_at | - | - |
| Mouse Expression Set 430 | 1415733_PM_a_at | - | - |
| Mouse Expression Set 430 | 1415733_a_at | - | - |
| Murine Genome U74v2 Set | 95409_at | - | - |
| Murine Genome U74v2 Set | 95409_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415733_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415733_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415733_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415733_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415733_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415733_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 737177 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251853 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251853 | - | - |

Tomm5
Genomic Map