Assay Details
Gene Symbol: 4930548J01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045462.1 | 3 | 638 | 126 | ||
GenBank mRNA | AK019751.1 | - | 3 | 639 | 126 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930470H14 gene | 4930470H14Rik | -36.5kb | Sense | Chr. 17 - 4044658 - 4082995 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 4930517M08 gene | 4930517M08Rik | 515.1kb | Anti-sense | Chr. 17 - 4634980 - 4637478 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783679 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08702288 | - | - |
Whole Mouse Genome G4122A | A_52_P129056 | AK007199.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P129056 | AK019751.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009219 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009304 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009199 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13285184 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009233 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009284 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009292 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009226 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009184 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13414536 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13119137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13423052 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009239 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009307 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13512094 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009294 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009223 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009208 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13263170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009251 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009172 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009303 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009273 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009281 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009214 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009249 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009262 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13423055 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009216 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009271 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009194 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13269550 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13059569 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009232 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009313 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13481534 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009187 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13097994 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009220 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009306 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009295 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13458396 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13409682 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009205 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009227 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009267 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009256 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13409681 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009291 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009180 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009312 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009242 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009274 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009221 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009268 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009275 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009261 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009315 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009277 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009212 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009186 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13119131 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009270 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009224 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009204 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009244 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009210 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009254 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009183 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009231 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009257 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009250 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009279 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009245 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009288 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13119136 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13269553 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009201 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 104590095 | - | - |
