Assay Details
| Gene Symbol: Tgfbrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013025.2 | NP_001013043.1 | 3 | 1086 | 78 | |
| GenBank mRNA | AK051333.1 | - | 2 | 2321 | 78 | |
| AK141742.1 | - | 2 | 642 | 78 | ||
| AK159332.1 | - | 2 | 619 | 78 | ||
| BC072608.1 | - | 3 | 1086 | 78 | ||
| BC138866.1 | - | 3 | 1000 | 8861888 | 78 | |
| BC138867.1 | - | 3 | 1000 | 8861889 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tgfbrap1
Genomic Map