Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400000733.mm.1 |
NM_028226.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000733.mm.1 |
NM_028226.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000733.mm.1 |
TC0400000108.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000740.mm.1 |
NM_028226.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000740.mm.1 |
NM_028226.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000740.mm.1 |
TC0400000108.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000737.mm.1 |
NM_028226.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000737.mm.1 |
NM_028226.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000737.mm.1 |
TC0400000108.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000739.mm.1 |
NM_028226.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000739.mm.1 |
NM_028226.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000739.mm.1 |
TC0400000108.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000741.mm.1 |
NM_028226.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000741.mm.1 |
NM_028226.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000741.mm.1 |
TC0400000108.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000734.mm.1 |
NM_028226.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000734.mm.1 |
NM_028226.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000734.mm.1 |
TC0400000108.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000735.mm.1 |
NM_028226.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000735.mm.1 |
NM_028226.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000735.mm.1 |
TC0400000108.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000730.mm.1 |
NM_028226.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000730.mm.1 |
NM_028226.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000730.mm.1 |
TC0400000108.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000736.mm.1 |
NM_028226.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000736.mm.1 |
NM_028226.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000736.mm.1 |
TC0400000108.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000738.mm.1 |
NM_028226.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000738.mm.1 |
NM_028226.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000738.mm.1 |
TC0400000108.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000742.mm.1 |
NM_028226.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000742.mm.1 |
NM_028226.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000742.mm.1 |
TC0400000108.mm.1 |
MTA TC |