Assay Details
| Gene Symbol: C230091D08Rik | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015479.1 | 2 | 1848 | 116 | |||
| GenBank mRNA | AK049008.1 | - | LIT3431 UBE3A-ATS |
2 | 1848 | 116 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C230091D08Rik
Genomic Map