Assay Details
| Gene Symbol: C030013G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_077216.1 | 3 | 474 | 134 | ||
| NR_077217.1 | 2 | 457 | 134 | |||
| GenBank mRNA | AK021075.1 | - | 2 | 459 | 134 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C030013G03Rik
Genomic Map