Assay Details
| Gene Symbol: 4933401L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016606.1 | - | 2 | 1237 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 31503 |
Gm31503 | -43.9kb | Sense | Chr. 18 - 83363538 - 83373741 on NCBI Build GRCm38 | |
| Downstream | teashirt zinc finger family member 1 | Tshz1 |
5730407I04Rik, D18Bwg1409e, Mtsh1, NY-CO-33, Sdccag33, Tsh1, mKIAA4206, teashirt1 |
593.2kb | Sense | Chr. 18 - 84011627 - 84086562 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933401L05Rik
Genomic Map