Assay Details
Gene Symbol: Txnl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025299.3 | NP_079575.1 | 4 | 82 | ||
GenBank mRNA | AK166497.1 | - | 5 | 82 | ||
BC022582.1 | - | 1 | 82 | |||
BC057058.1 | - | 1 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
